Order Entry
United States
ContactUsLinkComponent
Human Recombinant CRP (Prokaryotic) (from E. coli)
Human Recombinant CRP (Prokaryotic) (from E. coli)
Catalog # MSPP-RPA821HU1
Supplier:  Cloud-Clone
CAS Number:  
undefined
Human Recombinant CRP (Prokaryotic) (from E. coli)
Catalog # MSPP-RPA821HU1
Supplier:  Cloud-Clone
Supplier Number:  RPA821HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    1401
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    C Reactive Protein
  • UniProtKB:
    P02741
  • Protein/Peptide Name:
    CRP
  • Purity:
    90 - 100%
  • Molecular Weight:
    27 kDa
  • Sequence:
    Phe17~Pro224
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA821HU1

Specifications

About this item

This is a CRP recombinant protein (prokaryotic), Human is sequencing from pH e17~Pro224 with 90 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

C-reactive protein is a liver-derived pattern recognition molecule that is increased in inflammatory states. It rapidly increases within hours after tissue injury, and it is suggested that it is part of the innate immune system and contributes to host defense. Since cardiovascular disease is at least in part an inflammatory process, CRP has been investigated in the context of arteriosclerosis and subsequent vascular disorders. Based on multiple epidemiological and intervention studies, minor CRP elevation has been shown to be associated with future major cardiovascular risk. CRP is known for several decades as a nonspecific inflammation marker. High CRP levels are detected in human blood during bacterial, viral and other infections, as well as in noninfectious diseases such as rheumatic disorders and malignancies.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.