Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 µg
- Tag sequence:MGHHHHHHSGSEF
- Storage Conditions:–20 °C
- Endotoxin Content:<1.0 EU per 1 μg (determined by the LAL method)
- Gene ID:6441
- Reconstitution Instructions:Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Surfactant Associated Protein D
- UniProtKB:P35247
- Protein/Peptide Name:SPD
- Purity:80 - 100%
- Molecular Weight:43 kDa
- Sequence:Ala21~Phe375
- Endotoxin Level:Low
- Concentration:0.2 mg/ml
- Formulation:Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
- Nuclease-free:N
- Shipping Temperature:4 °C
- Tested Applications:Positive control, Immunogen, SDS-PAGE, Western blot.
- Cat. No.:MSPP-RPB039HU1
Specifications
About this item
This is a SPD recombinant protein (prokaryotic), Human is sequencing from Ala21~pH e375 with 80 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
- High quality, purity, reproducibility and effectiveness
- Offers customized buffers and tag options
- 100% quality and service satisfaction guarantee
Surfactant protein D (SP-D) belongs to the collectin family.These proteins are oligomeric proteins composed of carbohydrate-recognition domains(CRD)attached to collagenous regions.Collectins are structurally similar to the ficolins although they make use of different CRD structures:C-type lectin domain for the collectins.The lung is the major site of synthesis of SP-D, where the molecules are produced and secreted onto the epithelial surface by alveolar type II cells and unciliated bronchial epithelial cells. SP-D is also found in different epithelial cells of the gastrointestinal tract and in epithelial cells of various exocrine glands. SP-D is an important factor in the pulmonary anti-microbial defense. The anti-microbial defense mechanisms of SP-D are direct opsonization, neutralization and agglutination.