Order Entry
United States
ContactUsLinkComponent
Human Recombinant VEGFA (Prokaryotic) (from E. coli)
Human Recombinant VEGFA (Prokaryotic) (from E. coli)
Catalog # MSPP-RPA143HU1
Supplier:  Cloud-Clone
CAS Number:  
undefined
Human Recombinant VEGFA (Prokaryotic) (from E. coli)
Catalog # MSPP-RPA143HU1
Supplier:  Cloud-Clone
Supplier Number:  RPA143HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    7422
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Vascular Endothelial Growth Factor A
  • UniProtKB:
    P15692
  • Protein/Peptide Name:
    VEGFA
  • Purity:
    97 - 100%
  • Molecular Weight:
    17/18 kDa
  • Sequence:
    Pro28~Arg147
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA143HU1

Specifications

About this item

This is a VEGFA recombinant protein (prokaryotic), Human is sequencing from Pro28~Arg147 with 97 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

VEGF-A is a glycosylated mitogen that specifically acts on endothelial cells and has various effects, including mediating increased vascular permeability, inducing angiogenesis, vasculogenesis and endothelial cell growth, promoting cell migration, and inhibiting apoptosis. Alternatively spliced transcript variants, encoding either freely secreted or cell-associated isoforms, have been characterized. VEGF is a chemical signal produced by cells that stimulates the growth of new blood vessels. The most important member is VEGF-A. Other members are Placenta growth factor (PlGF), VEGF-B, VEGF-C and VEGF-D. The latter ones were discovered later than VEGF-A, and, before their discovery, VEGF-A was called just VEGF.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.