- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 µg
- Tag sequence:MGHHHHHHSGSEF
- Storage Conditions:–20 °C
- Endotoxin Content:<1.0 EU per 1 μg (determined by the LAL method)
- Gene ID:2099
- Reconstitution Instructions:Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Estrogen Receptor Alpha
- UniProtKB:P03372
- Protein/Peptide Name:ERa
- Purity:90 - 100%
- Molecular Weight:33 kDa
- Sequence:Ser178~Met438
- Endotoxin Level:Low
- Concentration:0.2 mg/ml
- Formulation:Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
- Nuclease-free:N
- Shipping Temperature:4 °C
- Tested Applications:Positive control, Immunogen, SDS-PAGE, Western blot.
- Cat. No.:MSPP-RPB050HU1
This is a ERa recombinant protein (prokaryotic), Human is sequencing from Ser178~Met438 with 90 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
- High quality, purity, reproducibility and effectiveness
- Offers customized buffers and tag options
- 100% quality and service satisfaction guarantee
Estrogen receptor structure-function is a vast topic and the subject of very active current research. Nuclear receptors are a large family of structurally related ligand-inducible transcription factors, including steroid receptors (SRs), thyroid/retinoids receptors (TR, RARs and RXRs), vitamin D receptors (VDR), LXR, PPARs, estrogen receptors (ERα and ERβ), and orphan receptors for which no ligand has been yet identified. While having in common a modular structure, they are activated by distinct lipophilic small molecules such as glucocorticoids, progesterone, estrogens, retinoids, and fatty acid derivatives.All nuclear receptors have a hydrophobic pocket into which its specific ligand binds, with helix 12 (H12) being the key response element of NRs.