- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 µg
- Tag sequence:MGHHHHHHSGSEF
- Storage Conditions:–20 °C
- Endotoxin Content:<1.0 EU per 1 μg (determined by the LAL method)
- Gene ID:445
- Reconstitution Instructions:Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Argininosuccinate Synthetase 1
- UniProtKB:P00966
- Protein/Peptide Name:ASS1
- Purity:90 - 100%
- Molecular Weight:47 kDa
- Sequence:Met1~Lys412
- Endotoxin Level:Low
- Concentration:0.2 mg/ml
- Formulation:Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
- Nuclease-free:N
- Shipping Temperature:4 °C
- Tested Applications:Positive control, Immunogen, SDS-PAGE, Western blot.
- Cat. No.:MSPP-RPC317HU1
This is a ASS1 recombinant protein (prokaryotic), Human is sequencing from Met1~Lys412 with 90 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
- High quality, purity, reproducibility and effectiveness
- Offers customized buffers and tag options
- 100% quality and service satisfaction guarantee
Argininosuccinate synthetase 1 catalyzes the penultimate step of the arginine biosynthetic pathway. There are approximately 10 to 14 copies of this gene including the pseudogenes scattered across the human genome, among which the one located on chromosome 9 appears to be the only functional gene for argininosuccinate synthetase. The ASS1 gene encodes ardininosuccinate synthetase-1, a cytosolic urea cycle enzyme mainly expressed in periportal hepatocytes, but also in most other body tissues. The enzyme is a homotetrameric protein composed of 45-kDa monomers and is involved in the synthesis of arginine and catalyzes that condensation of citrulline and aspartate to argininosuccinate using ATP.