Order Entry
United States
ContactUsLinkComponent
Rat Recombinant NGF (Prokaryotic) (from E. coli)
Rat Recombinant NGF (Prokaryotic) (from E. coli)
Catalog # MSPP-RPA105RA1
CAS Number:  
undefined
Rat Recombinant NGF (Prokaryotic) (from E. coli)
Catalog # MSPP-RPA105RA1
Supplier Number:  RPA105RA01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    310738
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Nerve Growth Factor
  • UniProtKB:
    P25427
  • Protein/Peptide Name:
    NGF
  • Purity:
    90 - 100%
  • Molecular Weight:
    33 kDa
  • Sequence:
    Glu19~Gly241
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA105RA1

Specifications

About this item

This is a NGF recombinant protein (prokaryotic), Rat is sequencing from Glu19~Gly241 with 90 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Nerve growth factor (NGF) is a naturally occurring molecule in the body which stimulates the growth and differentiation of the sympathetic and certain sensory nerves. NGF is a protein that consists of 3 types of polypeptide chains - alpha, beta and gamma that interact to form the protein. The NGF beta chain (NGFB) is solely responsible for the nerve growth stimulating activity of NGF. The NGFB gene is in chromosome band 1p22. An ointment containing NGF has been reported to help heal bedsores (decubiti).This complex contains 2 identical 118-amino acid beta-chains, which are solely responsible for nerve growth stimulating activity of NGF. Human DNA fragments coding for NGF were identified by Zabel et al. (1984) using a mouse submaxillary cDNA probe.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.