Order Entry
United States
ContactUsLinkComponent
Pierce™ Sulfhydryl Addition Kit, Thermo Scientific
Pierce™ Sulfhydryl Addition Kit, Thermo Scientific
Catalog # PI23460
Supplier:  Invitrogen
CAS Number:  
Pierce™ Sulfhydryl Addition Kit, Thermo Scientific
Catalog # PI23460
Supplier:  Invitrogen
Supplier Number:  23460
CAS Number:  

Specifications

  • Description:
    Pierce Sulfhydryl Addition Kit
  • Cat. No.:
    PI23460
  • No. of reactions:
    1
  • Supplier no.:
    23460

Specifications

About this item

Introduce free sulfhydryl groups into a protein of interest to allow the modified protein to react with a variety of sulfhydryl-selective products.

  • Adds a protected sulfhydryl that can be deprotected by hydroxylamine
  • Allows long-term storage of the sulfhydryl-modified molecule
  • Forms cleavable disulfide bonds with other sulfhydryl-containing molecules
  • Reacts with primary amines (e.g., lysine residues proteins) to form stable amide bonds
  • Preserves protein activity with its mild, non-denaturing reaction conditions

Sulfhydryl groups are added through a covalent linkage to yield a stable product, yet the reaction conditions will not denature the protein in order to preserve protein activity. In addition, the sulfhydryl group is protected to allow for long-term storage of the modified protein. Deprotection is accomplished through the use of a hydroxylamine, rather than a reducing agent to eliminate the presence of a second sulfhydryl that could quench or compete with subsequent reactions.

Delivery: The kit includes SATA (2 mg); Hydroxylamine-HCl (5 mg); 10X Conjugation Buffer stock (20 mL); Phosphate Buffered Saline pack (1 pack); Dimethylformamide (DMF, 1mL); Dextran desalting column (1x5 mL); Column extender (1); Ellman’s reagent (DTNB, 2 mg); and Cysteine-HCl H2O (20 mg).