Order Entry
United States
ContactUsLinkComponent
Human Recombinant AQP4 (prokaryotic)(from E. coli)
Human Recombinant AQP4 (prokaryotic)(from E. coli)
Catalog # MSPP-RPA582HU1
Supplier:  Cloud-Clone
CAS Number:  
undefined
Human Recombinant AQP4 (prokaryotic)(from E. coli)
Catalog # MSPP-RPA582HU1
Supplier:  Cloud-Clone
Supplier Number:  RPA582HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MSPILGYWKIKGLVQPTRLLLEYLEEKYEEHLYERDEGDKWRNKKFELGLEFPNLPYYIDGDVKLTQSMAIIRYIADKHNMLGGCPKERAEISMLEGAVLDIRYGVSRIAYSKDFETLKVDFLSKLPEMLKMFEDRLCHKTYLNGDHVTHPDFMLYDALDVVLYMDPMCLDAFPKLVCFKKRIEAIPQIDKYLKSSKYIAWPLQGWQATFGGGDHPPKSDGSTSGSGHHHHHHSAGLVPRGSTAIGMKETAAAKFERQHMDSPDLGTGGGSGIEGRGSMGYRGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    361
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Aquaporin 4
  • UniProtKB:
    P55087
  • Protein/Peptide Name:
    AQP4
  • Purity:
    90 - 100%
  • Molecular Weight:
    45 kDa
  • Sequence:
    Cys178~Gly317
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4 containing 0.01% SKL, 5% trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA582HU1

Specifications

About this item

This is a AQP4 recombinant protein (prokaryotic), Human is sequencing from Cys178~Gly317 with 90 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Aquaporin-4 (AQP4), is a member of the aquaporin family of intrinsic membrane proteins that function as water-selective channels in the plasma membranes of many cells. The encoded protein is the predominant aquaporin found in brain. Aquaporin-4 is located in many tissues, including the basolateral membrane of the renal cortical and medullary principal cell, as well as in the brain.Aquaporin-4 is the first specific molecule to be defined as a target for the autoimmune response in any form of multiple sclerosis. Aquaporin 4 is found in the basolateral cell membrane of principal collecting duct cells and provide a pathway for water to exit these cells.AQP4 is also expressed in astrocytes and is upregulated by direct insult to the central nervous system.Aquaporin-4 has been proposed as the primary autoimmune target in Devic's disease.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.