- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Size:50 µg
- Tag sequence:MGHHHHHHSGSEF
- Storage Conditions:–20 °C
- Endotoxin Content:<1.0 EU per 1 μg (determined by the LAL method)
- Gene ID:17700
- Reconstitution Instructions:Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Myostatin
- UniProtKB:O08689
- Protein/Peptide Name:MSTN
- Purity:95 - 100%
- Molecular Weight:15 kDa
- Sequence:Asp268~Ser376
- Endotoxin Level:Low
- Concentration:0.2 mg/ml
- Formulation:Lyophilized from PBS, pH 7.4 containing 0.01% SKL, 5% trehalose
- Nuclease-free:N
- Shipping Temperature:4 °C
- Tested Applications:Positive control, Immunogen, SDS-PAGE, Western blot.
- Cat. No.:MSPP-RPB653MU1
This is a MSTN recombinant protein (prokaryotic), Mouse is sequencing from Asp268~Ser376 with 95 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
- High quality, purity, reproducibility and effectiveness
- Offers customized buffers and tag options
- 100% quality and service satisfaction guarantee
Myostatin is a growth factor that limits muscle tissue growth i.e. higher concentrations of myostatin in the body may cause the individual to have less developed muscles. The myostatin protein is produced primarily in skeletal muscle cells, circulates in the blood and acts on muscle tissue, apparently by slowing down the development of muscle stem cells. The precise mechanism remains unknown. Its functions in non-mammalian vertebrates appear to be somewhat conserved as muscle-specific actions have been demonstrated in birds. However, it is produced in many different fish tissues, suggesting that it may regulate more than just muscle mass in these vertebrates. Human Myostatin consists of two identical subunits, each consisting of 110 amino acid residues. Its total molecular weight is 25.0 kDa.