- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Size:50 µg
- Tag sequence:MGHHHHHHSGSEF
- Storage Conditions:–20 °C
- Endotoxin Content:<1.0 EU per 1 μg (determined by the LAL method)
- Gene ID:17076
- Reconstitution Instructions:Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Lymphocyte Antigen 75
- UniProtKB:Q60767
- Protein/Peptide Name:LY75
- Purity:90 - 100%
- Molecular Weight:36 kDa
- Sequence:Cys216~Pro503
- Endotoxin Level:Low
- Concentration:0.2 mg/ml
- Formulation:Lyophilized from PBS, pH 7.4 containing 0.01% SKL, 5% trehalose
- Nuclease-free:N
- Shipping Temperature:4 °C
- Tested Applications:Positive control, Immunogen, SDS-PAGE, Western blot.
- Cat. No.:MSPP-RPB593MU1
This is a LY75 recombinant protein (prokaryotic), Mouse is sequencing from Cys216~Pro503 with 90 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
- High quality, purity, reproducibility and effectiveness
- Offers customized buffers and tag options
- 100% quality and service satisfaction guarantee
The LY75 protein has 77% overall amino acid identity with mouse Dec205. LY75 is a member of the MMR family of multidomain molecules. It contains a signal peptide, a cysteine-rich domain, a fibronectin type II domain, 10 carbohydrate recognition-like domains, a transmembrane domain, and a cytoplasmic tail. Northern blot analysis of hematopoietic cell lines revealed the presence of a predominant 7.8-kb and a secondary 9.5-kb transcript in myeloid, B-cell, and Hodgkin cell lines but not in the Jurkat T-cell line. In peripheral blood, immature DC contained barely detectable transcripts, which increased 50-fold upon differentiation and activation. LY75 acts as an endocytic receptor to direct captured antigens from the extracellular space to a specialized antigen-processing compartment.