Order Entry
United States
ContactUsLinkComponent
Bovine Recombinant CSN1 (prokaryotic)(from E. coli)
Bovine Recombinant CSN1 (prokaryotic)(from E. coli)
Catalog # MSPP-RPJ333BO1
CAS Number:  
undefined
Bovine Recombinant CSN1 (prokaryotic)(from E. coli)
Catalog # MSPP-RPJ333BO1
Supplier Number:  RPJ333BO01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Bovine
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    282208
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Casein Alpha
  • UniProtKB:
    P02662
  • Protein/Peptide Name:
    CSN1
  • Purity:
    97 - 100%
  • Molecular Weight:
    30 kDa
  • Sequence:
    Arg16~Gln187
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4 containing 0.01% SKL, 5% trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPJ333BO1

Specifications

About this item

This is a CSN1 recombinant protein (prokaryotic), Bovine is sequencing from Arg16~Gln187 with 97 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Milk casein can be separated by urea starch electrophoresis into 3 regions, alpha, beta, and kappa casein. Alpha and beta variants are present in the human population. In Italy the frequency of the 2 alpha alleles was 0.908 and 0.092, 2 beta alleles had a frequency of 0.678 and 0.322. The human alpha-S1, beta-, and kappa-casein genes are closely linked and arranged in that order. By fluorescence in situ hybridization, they demonstrated that the casein gene family is localized to 4q21.1. the human 'locus' comprises at least 4 casein genes: 3 genes encoding calcium-sensitive, casein-like genes, and 1 kappa-casein gene, in the order alpha-s1--beta--alpha-s2--kappa. The approximate size of the human casein gene locus is 350 kb. The casein cluster is located within 700 kb of the albumin gene cluster, which is located on 4q13.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.