- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Multiple
- Size:50 µg
- Tag sequence:MGHHHHHHSGSEF
- Storage Conditions:−20 °C
- Endotoxin Content:<1.0 EU per 1 µg (determined by the LAL method)
- Gene ID:60
- Reconstitution Instructions:Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Beta Actin
- UniProtKB:P60709
- Protein/Peptide Name:ACTB (prokaryotic)
- Purity:90 - 100%
- Molecular Weight:44 kDa
- Sequence:Met1~Phe375
- Endotoxin Level:Low
- Concentration:0.2 mg/ml
- Formulation:Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
- Nuclease-free:N
- Shipping Temperature:4 °C
- Tested Applications:Positive control, Immunogen, SDS-PAGE, Western blot.
- Cat. No.:MSPP-RPB340MI1
This is a ACTB recombinant protein (prokaryotic), Hu/Mu/Rat/Gu/Dog/Pig/Bv/Sheep/Hr/Ck is sequencing from Met1~Phe375 with 90 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
- High quality, purity, reproducibility and effectiveness
- Offers customized buffers and tag options
- 100% quality and service satisfaction guarantee
Beta-actin (gene name ACTB) is one of the two nonmuscle cytoskeletal actins. Actins are highly conserved proteins that are involved in cell motility, structure and integrity. Alpha actins are a major constituent of the contractile apparatus. In a neoplastic cell line resulting from treatment of cultured human diploid fibroblasts with a chemical mutagen, Leavitt et al. (1982) observed a mutant form of beta actin. Toyama and Toyama (1984) isolated and characterized lines of KB cells resistant to cytochalasin B. They found that one resistant line had an alteration in beta-actin. Such cells bound less cytochalasin B than did parental KB cells. The authors suggested that the primary site of action of cytochalasin B on cell motility processes is beta-actin.