Order Entry
United States
ContactUsLinkComponent
Human Recombinant BANF1 (prokaryotic) (from E.coli)
Human Recombinant BANF1 (prokaryotic) (from E.coli)
Catalog # MSPP-RPC336HU1
CAS Number:  
undefined
Human Recombinant BANF1 (prokaryotic) (from E.coli)
Catalog # MSPP-RPC336HU1
Supplier Number:  RPC336HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <1.0 EU per 1 µg (determined by the LAL method)
  • Gene ID:
    8815
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Barrier To Autointegration Factor 1
  • UniProtKB:
    O75531
  • Protein/Peptide Name:
    BANF1 (prokaryotic)
  • Purity:
    95 - 100%
  • Molecular Weight:
    14 kDa
  • Sequence:
    Thr2~Leu89
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPC336HU1

Specifications

About this item

This is a BANF1 recombinant protein (prokaryotic), Human is sequencing from Thr2~Leu89 with 95 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Barrier-to-autointegration factor was identified by its ability to protect retroviruses from intramolecular integration and therefore promote intermolecular integration into the host cell genome. The endogenous function of the protein is unknown.

The protein forms a homodimer which localizes to the nucleus and is specifically associated with chromosomes during mitosis. This protein binds to DNA in a non-specific manner and studies in rodents suggest that it also binds to lamina-associated polypeptide 2, a component of the nuclear lamina. It also associates with the LEM Domain containing proteins LAP2, Emerin, and MAN1.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.