Order Entry
United States
ContactUsLinkComponent
Human Recombinant SLC30A8 (prokaryotic) (from E.coli)
Human Recombinant SLC30A8 (prokaryotic) (from E.coli)
Catalog # MSPP-RPE490HU2
Supplier:  Cloud-Clone
CAS Number:  
undefined
Human Recombinant SLC30A8 (prokaryotic) (from E.coli)
Catalog # MSPP-RPE490HU2
Supplier:  Cloud-Clone
Supplier Number:  RPE490HU02
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <1.0 EU per 1 µg (determined by the LAL method)
  • Gene ID:
    169026
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Solute Carrier Family 30 Member 8
  • UniProtKB:
    Q8IWU4
  • Protein/Peptide Name:
    SLC30A8 (prokaryotic)
  • Purity:
    97 - 100%
  • Molecular Weight:
    10/15 kDa
  • Sequence:
    Met1~Ile80
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPE490HU2

Specifications

About this item

This is a SLC30A8 recombinant protein (prokaryotic), Human is sequencing from Met1~Ile80 with 97 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

SLC30A8 is expressed at a high level only in the pancreas, particularly in islets of Langerhans. The protein colocalizes with insulin in the secretory pathway granules of the insulin-secreting INS-1 cells. Allelic variants of this gene exist that confer susceptibility to diabetes mellitus, noninsulin-dependent. Several transcript variants encoding different isoforms have been found for this gene.It was also expressed in fetal pancreas, islets of Langerhans, and an insulinoma cell line, but not in HeLa cells. Fluorescence-tagged ZNT8 was expressed in a punctate pattern in the cytoplasm of transfected insulinoma cells and HeLa cells. In insulinoma cells, ZNT8 expression overlapped that of insulin, suggesting that ZNT8 is associated with insulin granule secretion.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.