Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant ELA3B (prokaryotic) (from E.coli)
  MSPP-RPE212HU1
 :  
undefined
Human Recombinant ELA3B (prokaryotic) (from E.coli)
  MSPP-RPE212HU1
 :  RPE212HU01
 :  

 

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <1.0 EU per 1 µg (determined by the LAL method)
  • Gene ID:
    23436
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Elastase 3B
  • UniProtKB:
    P08861
  • Protein/Peptide Name:
    ELA3B (prokaryotic)
  • Purity:
    95 - 100%
  • Molecular Weight:
    27 kDa
  • Sequence:
    Val29~His270
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPE212HU1

 

 

This is a ELA3B recombinant protein (prokaryotic), Human is sequencing from Val29~His270 with 95 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Humans have six elastase genes which encode the structurally similar proteins elastase 1, 2, 2A, 2B, 3A, and 3B. Unlike other elastases, elastase 3B has little elastolytic activity. Like most of the human elastases, elastase 3B is secreted from the pancreas as a zymogen and, like other serine proteases such as trypsin, chymotrypsin and kallikrein, it has a digestive function in the intestine. Elastase 3B preferentially cleaves proteins after alanine residues. Elastase 3B may also function in the intestinal transport and metabolism of cholesterol. Both elastase 3A and elastase 3B have been referred to as protease E and as elastase 1, and excretion of this protein in fecal material is frequently used as a measure of pancreatic function in clinical assays..

 : ISO 9001 and ISO 13485 certified, three level QC system.
 : For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.