Order Entry
United States
ContactUsLinkComponent
Human Recombinant MMP9 (prokaryotic) (from E.coli)
Human Recombinant MMP9 (prokaryotic) (from E.coli)
Catalog # MSPP-RPA553HU1
Supplier:  Cloud-Clone
CAS Number:  
undefined
Human Recombinant MMP9 (prokaryotic) (from E.coli)
Catalog # MSPP-RPA553HU1
Supplier:  Cloud-Clone
Supplier Number:  RPA553HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <1.0 EU per 1 µg (determined by the LAL method)
  • Gene ID:
    4318
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Matrix Metalloproteinase 9
  • UniProtKB:
    P14780
  • Protein/Peptide Name:
    MMP9 (prokaryotic)
  • Purity:
    95 - 100%
  • Molecular Weight:
    20 kDa
  • Sequence:
    Gly213~Ala399
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA553HU1

Specifications

About this item

This is a MMP9 recombinant protein (prokaryotic), Human is sequencing from Gly213~Ala399 with 95 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

MMP-9 (also known as gelatinase B) is secreted as a 92 kDa zymogen. Cleavage of pro-MMP-9 at or near residue 87 results in the active enzyme with a mass of approximately 82 kDa. MMP-9 has three fibronectin type II domains, a hemopexin-like domain and a proline-rich type V collagen-homologous domain. Pro-MMP-9 can be activated by MMP-3 or by certain bacterial proteinases. MMP-9 is inhibited byα2-macroglobulin or by TIMP-1, which binds to pro-MMP-9 as well as to active MMP-9. Pro-MMP-9 is secreted by monocytes, macrophages, neutrophils, keratinocytes, fibroblasts, osteoclasts, chondrocytes, skeletal muscle satellite cells, endothelial cells, and various tumor cells. Pro-MMP-9 expression is upregulated by TGF-β1, IL-1β, TGF-α, PDGF-AB.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.