Order Entry
United States
ContactUsLinkComponent
Human Recombinant PIIINP (eukaryotic) (from 293Fcells)
Human Recombinant PIIINP (eukaryotic) (from 293Fcells)
Catalog # MSPP-EPA573H61
CAS Number:  
undefined
Human Recombinant PIIINP (eukaryotic) (from 293Fcells)
Catalog # MSPP-EPA573H61
Supplier Number:  EPA573HU61
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <1.0 EU per 1 µg (determined by the LAL method)
  • Gene ID:
    1281
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Procollagen III N-Terminal Propeptide
  • UniProtKB:
    P02461
  • Protein/Peptide Name:
    PIIINP (eukaryotic)
  • Purity:
    97 - 100%
  • Molecular Weight:
    30 kDa
  • Sequence:
    Gln24~Pro153
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 5% Trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-EPA573H61

Specifications

About this item

This is a PIIINP recombinant protein (eukaryotic,293F Cell), Human is sequencing from Gln24~Pro153 with 97 to 100% purity. Lyophilized from PBS, pH 7.4, containing 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Plasma levels of PIIINP a marker of extracellular matrix metabolism activity have been linked to abdominal aortic aneurysm growth rate, but its relationship with thoracic aortic aneurysm (TAA) growth has never previously been studied. The PIIINP molecule is an indicator of both the synthesis and the degradation of type III collagen. Occasionally, the removal of PIIINP from the newly-synthesized type III collagen is not complete and the propeptide may remain attached to some of the molecules, known as type IIIpN-collagen. PIIINP is a degradation product of type IIIpN collagen. PIIINP has a molecular weight of 42000 and contains three distinct domains: a triple-helical domain (Col 3) in the middle of the molecule, the Col 1-domain at the aminoterminal and Col 2-domain at the carboxyterminal end of the propeptide. PIIINP is cleared from the circulation by scavenger receptors in liver endothelial cells.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.