Order Entry
United States
ContactUsLinkComponent
 
Mouse Recombinant GPX4 (prokaryotic) (from E.coli)
Catalog # MSPP-RPC994MU1
CAS Number:  
undefined
Mouse Recombinant GPX4 (prokaryotic) (from E.coli)
Catalog # MSPP-RPC994MU1
Supplier Number:  RPC994MU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <1.0 EU per 1 µg (determined by the LAL method)
  • Gene ID:
    625249
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Glutathione Peroxidase 4
  • UniProtKB:
    O70325
  • Protein/Peptide Name:
    GPX4 (prokaryotic)
  • Purity:
    90 - 100%
  • Molecular Weight:
    18 kDa
  • Sequence:
    Gly74~Leu197
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPC994MU1

Specifications

About this item

This is a GPX4 recombinant protein (prokaryotic), Mouse is sequencing from Gly74~Leu197 with 90 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

The antioxidant enzyme (GPx4) belongs to the family of glutathione peroxidases, which consists of 8 known mammalian isoenzymes (GPx1-8). Gpx4 catalyzes the reduction of hydrogen peroxide, organic hydroperoxides, and lipid peroxides at the expense of reduced glutathione and functions in the protection of cells against oxidative stress. The oxidized form of glutathione (glutathione disulfide), which is generated during the reduction of hydroperoxides by GPx4, is recycled by glutathione reductase and NADPH/H+. GPx4 differs from the other GPx family members in terms of its monomeric structure, a less restricted dependence on glutathione as reducing substrate, and the ability to reduce lipid-hydroperoxides inside biological membranes.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.