Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant C3 (prokaryotic) (from E.coli)
Mouse Recombinant C3 (prokaryotic) (from E.coli)
Catalog # MSPP-RPA861MU1
CAS Number:  
undefined
Mouse Recombinant C3 (prokaryotic) (from E.coli)
Catalog # MSPP-RPA861MU1
Supplier Number:  RPA861MU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <1.0 EU per 1 µg (determined by the LAL method)
  • Gene ID:
    12266
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Complement Component 3
  • UniProtKB:
    P01027
  • Protein/Peptide Name:
    C3 (prokaryotic)
  • Purity:
    90 - 100%
  • Molecular Weight:
    42 kDa
  • Sequence:
    Ala965~Arg1303
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA861MU1

Specifications

About this item

This is a C3 recombinant protein (prokaryotic), Mouse is sequencing from Ala965~Arg1303 with 90 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Complement C3 is the major plasma proteins of the immune system complement pathways. The synthesis of these proteins is increased in response to inflammation and infection but at a slower rate than for traditional acute phase proteins. Complement protein C3 is the fourth component to attach in the complement reaction sequence. It is a beta-globulin with a sedimentation coefficient of 5.5 and a molecular weight of 185000. C3 is a central molecule in the complement system whose activation is essential for all the important functions performed by this system. C3 can promote phagocytosis during an inflammatory response against pathogens, but unregulated activation of C3 could lead to host cell damage. Low level of serum C3 associates with hypocomplementaemia, primary biliary cirrhosis

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.