Order Entry
United States
ContactUsLinkComponent
Human Recombinant PINP (prokaryotic) (from E.coli)
Human Recombinant PINP (prokaryotic) (from E.coli)
Catalog # MSPP-RPA957HU1
CAS Number:  
undefined
Human Recombinant PINP (prokaryotic) (from E.coli)
Catalog # MSPP-RPA957HU1
Supplier Number:  RPA957HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGSSHHHHHHSSGHMASMSDSEVNQEAKPEVKPEVKPETHINLKVSDGSSEIFFKIKKTTPLRRLMEAFAKRQGKEMDSLRFLYDGIRIQADQTPEDLDMEDNDIIEAHREQIGLVPRGSGSEF
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <1.0 EU per 1 µg (determined by the LAL method)
  • Gene ID:
    1277
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Procollagen I N-Terminal Propeptide
  • UniProtKB:
    P02452
  • Protein/Peptide Name:
    PINP (prokaryotic)
  • Purity:
    95 - 100%
  • Molecular Weight:
    40 kDa
  • Sequence:
    Gln23~Pro161
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA957HU1

Specifications

About this item

This is a PINP recombinant protein (prokaryotic), Human is sequencing from Gln23~Pro161 with 95 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

The PINP molecule is similar to PIIINP consisting of three distinct structural domains: Col 1 is on the aminoterminal side of the molecule, while Col 2 and Col 3 are situated on the middle of the helically structured molecule (Kühn et al. 1982). The PINP molecule has a molecular mass of 35 000 and is cleared by scavenger receptors in liver endothelial cells (Melkko et al. 1994). PINP often occurs in circulation in two forms of different molecular sizes. One is identical to the trimeric authentic antigen (intact PINP) whereas the other consists of smaller forms of PINP, resembling a single domain of the proα1(I) chain of PINP and is probably a degradation product of type I procollagen or I pN-collagen. Thus, an assay of intact PINP rather than total PINP appears to be more sensitive in detecting changes in the rate of type I collagen synthesis (Melkko et al. 1996, Risteli and Risteli 1999).

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.