ContactUsLinkComponent
Ward's® Triple Instrument Bundle
Ward's® Triple Instrument Bundle
Catalog # 470357-320
New Product
undefined
Ward's® Triple Instrument Bundle
Catalog # 470357-320
Supplier Number:  121008
New Product
Specifications
  • Description:
    Triple instrument bundle
  • Cat. No.:
    470357-320
Specifications
About this item
Spin-down, shake, and stir with this all-in-one equipment bundle.
  • Get the full functionality of three machines in two pieces of equipment!
  • Complete with a Ward’s® Silicone Lab Mat to protect your workspace from spills and wear
Ward’s® Magfuge® centrifuge/stirrer (470316-832) - The industries first high-speed centrifuge (12,500 rpm) and magnetic stirrer (3 L), combined into one unit. Offering durability, chemical resistance, and easy maintenance. It is compact, low-profile design saves bench space and reduces lab clutter.
Mini vortexer (70316-814) - Small footprint Vortexer offering maximum vortex shaking for a variety of tube sizes. A simple touch start operation allows for immediate sample mixing with one hand and automatic stop when tube is lifted.
Ward’s® Silicone lab mat (470316-832) - This FDA food-grade silicone lab mat is ideal for high sterility
environments, protecting counter tops from temperatures up to 200 °C (392 °F) and resisting moderate oxidizing chemicals.