About this item
The mouse monoclonal antibody M19H specifically binds to the scFv region of a CD19-specific mouse monoclonal antibody (mAb, clone FMC63). CD19 antigen is a B-cell specific cell surface antigen, which is expressed in all B-cell lineage malignancies and normal B-cells. The scFv region of FMC63 has been used to develop CD19-specific chimeric antigen receptor (CAR) T cells utilized in clinical trials.
- High sensitivity
- Low non-specificity
- Idiotypic
- ADA capable
- Diagnostic material for CDx
Reliable CAR-T CDx, been in helping commercialized CAR-T therapeutics. Immediate shipping if in stock, lead time might be up to 30 days if out of stock pending for reproduction.
Type: Primary
Antigen: fmc63 scFv
Clonality: Monoclonal
Clone: M19H
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1a kappa
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- MSPP-300419
- MSPP-300428
- MSPP-300427
- Antigen Symbol:
- fmc63 scFv
- fmc63 scFv
- fmc63 scFv
- Antigen Name:
- CD19 CAR, scFv region of a CD19-specific mouse mAb clone FMC63
- CD19 CAR, scFv region of a CD19-specific mouse mAb clone FMC63
- CD19 CAR, scFv region of a CD19-specific mouse mAb clone FMC63
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- M19H
- M19H
- M19H
- Reactivity:
- Mouse
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1a kappa
- IgG1a kappa
- IgG1a kappa
- Flow Cytometry:
- Yes
- Yes
- Yes
- Immunogen:
- scFv region of a CD19-specific mouse mAb clone FMC63
- scFv region of a CD19-specific mouse mAb clone FMC63
- scFv region of a CD19-specific mouse mAb clone FMC63
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Protein A affinity purification
- Concentration:
- 1.0 mg/ml
- 1.0 mg/ml
- 1.0 mg/ml
- Storage Buffer:
- PBS, pH 7.4
- PBS, pH 7.4
- PBS, pH 7.4
- Storage Temperature:
- At −20 °C
- At −20 °C
- At −20 °C
- Shipping Temperature:
- 20
- 20
- 20