About this item
The rabbit monoclonal antibody R19M specifically binds to the scFv region of a CD19-specific mouse monoclonal antibody (mAb, clone FMC63). CD19 antigen is a B-cell specific cell surface antigen, which is expressed in all B-cell lineage malignancies and normal B-cells. The scFv region of FMC63 has been used to develop CD19-specific chimeric antigen receptor (CAR) T cells utilized in clinical trials.
- High sensitivity
- Low non-specificity
- Idiotypic
- Diagnostic material for CAR-T CDx
Reliable CAR-T CDx, been in helping commercialized CAR-T therapeutics. Immediate shipping if in stock, lead time might be up to 30 days if out of stock pending for reproduction.
Type: Primary
Antigen: fmc63 scFv
Clonality: Monoclonal
Clone: R19M
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- MSPP-200123
- MSPP-200124
- MSPP-200153
- MSPP-200154
- Antigen Symbol:
- fmc63 scFv
- fmc63 scFv
- fmc63 scFv
- fmc63 scFv
- Antigen Name:
- CD19 CAR, scFv region of a CD19-specific mouse mAb clone FMC63
- CD19 CAR, scFv region of a CD19-specific mouse mAb clone FMC63
- CD19 CAR, scFv region of a CD19-specific mouse mAb clone FMC63
- CD19 CAR, scFv region of a CD19-specific mouse mAb clone FMC63
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- R19M
- R19M
- R19M
- R19M
- Reactivity:
- Mouse
- Mouse
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- IgG
- Flow Cytometry:
- Yes
- Yes
- Yes
- Yes
- Immunogen:
- scFv region of a CD19-specific mouse mAb clone FMC63
- scFv region of a CD19-specific mouse mAb clone FMC63
- scFv region of a CD19-specific mouse mAb clone FMC63
- scFv region of a CD19-specific mouse mAb clone FMC63
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Protein A affinity purification
- Protein A affinity purification
- Concentration:
- 5 μl per test, recommended
- 5 μl per test, recommended
- 5 μl per test, recommended
- 5 μl per test, recommended
- Storage Buffer:
- Aqueous buffered solution containing protein stabilizer and ≤0.05% ProClin 300
- Aqueous buffered solution containing protein stabilizer and ≤0.05% ProClin 300
- Aqueous buffered solution containing protein stabilizer and ≤0.05% ProClin 300
- Aqueous buffered solution containing protein stabilizer and ≤0.05% ProClin 300
- Storage Temperature:
- At 2...8 °C, in dark place
- At 2...8 °C, in dark place
- At 2...8 °C, in dark place
- At 2...8 °C, in dark place
- Shipping Temperature:
- 20
- 20
- 20
- 20