Order Entry
United States
ContactUsLinkComponent
Anti-2C6 scFv Rabbit Monoclonal Antibody [clone: 1F5] (PE (Phycoerythrin))
Anti-2C6 scFv Rabbit Monoclonal Antibody [clone: 1F5] (PE (Phycoerythrin))
Supplier:  CYTOART, INC. MS
Certificates
Anti-2C6 scFv Rabbit Monoclonal Antibody [clone: 1F5] (PE (Phycoerythrin))
Supplier:  CYTOART, INC. MS
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • MSPP-202705
  • MSPP-202706
  • Antigen Symbol:
  • 2C6 scFv
  • 2C6 scFv
  • Antigen Name:
  • CD20 CAR, scFv region of a CD20-specific Human mAb clone 2C6
  • CD20 CAR, scFv region of a CD20-specific Human mAb clone 2C6
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • 1F5
  • 1F5
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG1
  • IgG1
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • scFv region of a CD20-specific Human mAb clone 2C6
  • scFv region of a CD20-specific Human mAb clone 2C6
  • Purification:
  • Protein A affinity purification
  • Protein A affinity purification
  • Concentration:
  • 1 μl per test, recommended
  • 1 μl per test, recommended
  • Storage Buffer:
  • Aqueous buffered solution containing protein stabilizer and ≤0.05% ProClin 300
  • Aqueous buffered solution containing protein stabilizer and ≤0.05% ProClin 300
  • Storage Temperature:
  • At 2...8 °C, in dark place
  • At 2...8 °C, in dark place
  • Shipping Temperature:
  • 20
  • 20

Specifications

Frequently Bought Together