
1-Benzyl-3-methylimidazolium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Certificates
About this item
CAS Number:500996-04-3
:
Formula:C₁₁H₁₃BF₄N₂
MDL Number:MFCD07784449
:260.04
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:60.0 to 64.0 deg-C
Specification Test Results
Frequently Bought Together