ContactUsLinkComponent
Clean-Ceil™ Fan Filter Modules, Microzone
Catalog # 35995-762
Supplier:  Avantor
Clean-Ceil™ Fan Filter Modules, Microzone
Catalog # 35995-762
Supplier:  Avantor
Specifications
  • Description:
    Clean-Ceil™ Fan Filter Module
  • Cat. No.:
    35995-762
  • Width:
    5 ft.
  • Supplier no.:
    FFM-2-4-A
Specifications
About this item
Ideal for use in cleanroom and sterile suite ceiling systems
These terminal fan filter HEPA units may be incorporated into existing facilities as a retrofit to upgrade cleanliness levels, or may be integrated into the design of new cleanroom installations. The energy-efficient Clean-Ceil™ filter is also ideal for use in modular or portable cleanrooms. Standard filter module comes complete with removable, washable, 40% efficient ASHRAE-rated prefilter and solid-state speed control.