ContactUsLinkComponent
Water-Jel® Fire Blanket and Heat Shield, Honeywell Safety
Water-Jel® Fire Blanket and Heat Shield, Honeywell Safety
Catalog # 56613-226
Supplier:  Avantor
undefined
Water-Jel® Fire Blanket and Heat Shield, Honeywell Safety
Catalog # 56613-226
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Mounting Bracket for Fire Blanket
  • Environmentally Preferable:
  • Cat. No.:
    56613-226
  • Supplier no.:
    049010
Specifications
About this item
Unique, simple-to-use products for emergency burn care do not require a water source or special training.
Standard size blanket is used to extinguish flames on a burning victim. Larger heat shield protects rescuer from heat and flames and extinguishes flames on victim. Each size comes in handy canister.
Can be used for all types of burns. When applied, blanket immediately cools the skin, eases pain, helps calm the victim and protects the wound from further contamination.