PYREX® Measuring (Mohr) Pipettes, Color-Coded, Tempered Top and Tip, Colored Markings, Corning
Supplier: Corning
Certificates
About this item
These reusable glass pipettes are color-coded by size for easy identification and sorting.
- Top end is constricted (Mohr type)
- Colored graduations are enamelled onto the glass
- Calibrated 'to deliver' (TD) their total capacity without blow-out
Sharp, fine lines provide accurate reading. Large numbers are easy to read. The top end of 5 to 25 ml sizes is constricted.
Certifications: ASTM E-1293.
Specifications
- Catalog No:
- 53083-119
- 53083-072
- 53083-050
- 53083-120
- 53083-039
- 53083-094
- 53083-006
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental Certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey