Specifications
- Antibody Type:Primary
- Antigen Name:Mucin 3
- Antigen Symbol:MUC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:MUC3A|Mucin 3A,Intestinal
- Amino Acid Number:1 - 101
- UniProtKB:Q3U5A2
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:15 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.45 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant MUC3(Asp1~Arg101)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAB031MU1
- Packaging:Vial
Specifications
About this item
Polyclonal antibody to Mucin 3 (MUC3), derived from recombinant MUC3(Asp1~Arg101), is reactive with Mouse.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee!
The mucin genes encode epithelial glycoproteins, some of which are secreted and some membrane bound. Each of the genes contains at least one large domain of tandemly repeated sequence that encodes the peptide sequence rich in serine and/or threonine residues, which carries most of the O-linked glycosylation. MUC3 gene, demonstrating that it encodes a transmembrane mucin and is subject to alternative splicing to a form lacking 1 EGF-like domain and the transmembrane domain. Structural homology of the human MUC3 sequence with that of rat and mouse Muc3 indicated that it is closely related to the rodent proteins and is likely to be involved in ligand binding and intracellular signaling.
Type: Primary
Antigen: MUC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse