Specifications
- Antibody Type:Primary
- Antigen Name:Triadin
- Antigen Symbol:TRDN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:59299
- Antigen Synonyms:TRISK|TDN|TRISK51
- Amino Acid Number:486 - 687
- UniProtKB:Q9QX75
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:30 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant TRDN(Leu486~Gln687)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAD033RA1
- Packaging:Vial
Specifications
About this item
Polyclonal antibody to Triadin (TRDN), derived from recombinant TRDN(Leu486~Gln687), is reactive with Rat.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee!
Triadin also known as TRDN, associated with the release of calcium ions from the sarcoplasmic reticulum triggering muscular contraction through calcium-induced calcium release.,Triadin, a protein found in rabbit triadic junctions, is intrinsic to the terminal cisternae and is closely associated with the RYR1. The predicted 729-amino acid human protein shares 95% identity with rabbit triadin. Like the rabbit protein, human triadin contains a small cytoplasmic domain and a single transmembrane domain. Human triadin has a calculated pI of 9.3 and molecular mass of 82 kDa. However, on Western blots, its apparent molecular mass is 117 kDa.
Type: Primary
Antigen: TRDN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat