- Antibody Type:Primary
- Antigen Name:Ferritin, Heavy Polypeptide
- Antigen Symbol:FTH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:2495
- Antigen Synonyms:PLIF|FTH1|PIG15|Apoferritin|Placenta Immunoregulatory Factor|Proliferation-Inducing Protein 15|Cell proliferation-inducing gene 15 protein|Ferritin Heavy Chain|FTHL6
- Amino Acid Number:1 - 183
- UniProtKB:P02794
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:25 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant FTH(Met1~Ser183)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAD021HU1
- Packaging:Vial
Polyclonal antibody to Ferritin, Heavy Polypeptide (FTH), derived from recombinant FTH(Met1~Ser183), is reactive with Human/Mouse.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee!
Ferritin heavy chain is a heavy subunit of ferritin, the major intracellular iron storage protein in prokaryotes and eukaryotes. It is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in ferritin proteins are associated with several neurodegenerative diseases. This gene has multiple pseudogenes. Several alternatively spliced transcript variants have been observed, but their biological validity has not been determined. The functionality of the gene was demonstrated by the fact that both transient transfectants and stable transformants of mouse fibroblasts actively transcribed human ferritin heavy-chain mRNA.
Type: Primary
Antigen: FTH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse