- Antibody Type:Primary
- Antigen Name:Semaphorin 5B
- Antigen Symbol:SEMA5B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:54437
- Antigen Synonyms:SemG|Sema Domain,Immunoglobulin Domain(Ig),Transmembrane Domain(TM)and Short Cytoplasmic Domain 5B|SEMAG
- Amino Acid Number:36 - 161
- UniProtKB:Q9P283
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:14 kDa
- Storage Temperature:Store at 2...8 °C for frequent use,−20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:recombinant SEMA5B (Leu36~Glu161)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAL926HU1
- Packaging:Vial
Polyclonal Antibody to Semaphorin 5B (SEMA5B), derived from recombinant SEMA5B (Leu36~Glu161), is reactive with Human/Rat.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Members of the semaphorin protein family, such as SEMA5B, are involved in axonal guidance during neural development. The deduced 1,093-amino acid protein contains an N-terminal signal sequence, followed by a semaphorin domain, 7 thrombospondin type-1 repeats, a putative transmembrane domain, and a short cytoplasmic tail. It also has 13 conserved cysteines and a conserved N-glycosylation site. Northern blot analysis of whole mouse embryos detected Semg at embryonic day 10, and its expression decreased toward birth. During development, Semg was expressed exclusively in the neuroepithelium along the entire anterioposterior axis. In adult animals, Semg was detected only in brain. The deduced 1,202-amino acid protein shares 93% identity with mouse Semg.
Type: Primary
Antigen: SEMA5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat