- Antibody Type:Primary
- Antigen Name:Lipopolysaccharide
- Antigen Symbol:LPS
- Clonality:Monoclonal
- Clone:C9
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Multiple
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:Lipo-Oligosaccharide|Lipoglycans|LOS|Lipooligosaccharide|Endotoxin
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Storage Temperature:Store at 2...8 °C for frequent use,−20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:OVA conjugated LPS
- Purification:Protein A + Protein G affinity chromatography
- Cat. No.:MSPP-LB526GE82
- Packaging:Vial
FITC-Linked Monoclonal Antibody to Lipopolysaccharide (LPS), derived from OVA conjugated LPS, is reactive with General species.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
LPS is a major component of the outer membrane of Gram-negative bacteria, contributing greatly to the structural integrity of the bacteria, and protecting the membrane from certain kinds of chemical attack. LPS also increases the negative charge of the cell membrane and helps stabilize the overall membrane structure. It is of crucial importance to gram negative bacteria, whose death results if it is mutated or removed. LPS is an endotoxin, and induces a strong response from normal animal immune systems. LPS acts as the prototypical endotoxin because it binds the CD14/TLR4/MD2 receptor complex, which promotes the secretion of pro-inflammatory cytokines in many cell types, but especially in macrophages. In Immunology, the term 'LPS challenge' refers to the process of exposing a subject to an LPS which may act as a toxin.
Type: Primary
Antigen: LPS
Clonality: Monoclonal
Clone: C9
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: General species