- Antibody Type:Primary
- Antigen Name:Aspartate Aminotransferase
- Antigen Symbol:AST
- Clonality:Monoclonal
- Clone:C1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Pig,Human,Rat,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:2805
- Antigen Synonyms:Glutamic-Oxaloacetic Transaminase 1,Soluble|AAT|Transaminase A|cCAT|Cysteine transaminase, cytoplasmic|SGOT|GOT1|ASAT|Aspartate Transaminase 1,Cytoplasmic
- Amino Acid Number:1 - 413
- UniProtKB:P17174
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:44 kDa
- Storage Temperature:Store at 2...8 °C for frequent use,−20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:recombinant AST (Met1~Gln413)
- Purification:Protein A + Protein G affinity chromatography
- Cat. No.:MSPP-MB214HU21
- Packaging:Vial
Monoclonal Antibody to Aspartate Aminotransferase (AST), derived from recombinant AST (Met1~Gln413), is reactive with Human/Mouse/Rat/Pig.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
AST) is similar to alanine transaminase (ALT) in that it is another enzyme associated with liver parenchymal cells. It is raised in acute liver damage. It is also present in red blood cells and cardiac muscle, skeletal muscle, and kidney and brain tissue, and may be elevated due to damage to those sources as well. AST was defined as a biochemical marker for the diagnosis of acute myocardial infarction in 1954. However the use of AST for such a diagnosis is now redundant and has been superseded by the cardiac troponins. The 2 isoenzymes are homodimeric. In liver about 80% of the enzyme activity is mitochondrial in origin, whereas in serum the enzyme activity is largely cytosolic.
Type: Primary
Antigen: AST
Clonality: Monoclonal
Clone: C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat, Pig