Specifications
- Antibody Type:Primary
- Antigen Name:Microtubule Associated Protein RP/EB Family, Member 1
- Antigen Symbol:MAPRE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Pig,Human
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:22919
- Antigen Synonyms:APC-binding protein EB1|Adenomatous Polyposis Coli-Binding protein|End-binding protein 1|EB1
- Amino Acid Number:2 - 268
- UniProtKB:Q15691
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:33.56 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant MAPRE1 (Ala2~Tyr268)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAG491HU1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Microtubule Associated Protein RP/EB Family, Member 1 (MAPRE1), derived from recombinant MAPRE1 (Ala2~Tyr268), is reactive with Human/Pig.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
MAPRE1 is a regulator of microtubule dynamics that localizes at both the growing plus ends of microtubules and the centrosome. It is involved in a variety of cellular processes, including establishment and maintenance of cell polarity, search and capture of chromosomes during mitosis, and positioning of the mitotic spindle during asymmetric cell division., This microtubule localization was abolished in the presence of the microtubule-destabilizing drug nocodazole; upon drug removal, the microtubule distribution of EB1 was recovered and EB1 fluorescence concentrated at the microtubule-organizing center. These results suggested that EB1 is associated with the microtubule network and may be involved in microtubule polymerization and spindle function.
Type: Primary
Antigen: MAPRE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Pig