Order Entry
United States
ContactUsLinkComponent
Anti-SIGIRR Rabbit Polyclonal Antibody
Anti-SIGIRR Rabbit Polyclonal Antibody
Catalog # MSPP-PAM229HU1
undefined
Anti-SIGIRR Rabbit Polyclonal Antibody
Catalog # MSPP-PAM229HU1
Supplier Number:  PAM229HU01

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Single Ig IL1 Related Receptor
  • Antigen Symbol:
    SIGIRR
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Pig,
    Human
  • Western Blot:
    Yes
  • Size:
    200 µl
  • Cross Adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    59307
  • Antigen Synonyms:
    Single Ig IL-1R-related molecule|Toll/interleukin-1 receptor 8|TIR8|Single immunoglobulin domain-containing IL1R-related protein
  • Amino Acid Number:
    1 - 118
  • UniProtKB:
    Q6IA17
  • Storage Buffer:
    PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
  • Molecular Weight:
    14 kDa
  • Storage Temperature:
    Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
  • Concentration:
    0.5 mg/ml (for reference only)
  • Shipping Temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant SIGIRR (Met1~His118)
  • Purification:
    Antigen-specific affinity chromatography followed by Protein A affinity chromatography
  • Cat. No.:
    MSPP-PAM229HU1
  • Packaging:
    Vial

Specifications

About this item

Polyclonal Antibody to Single Ig IL1 Related Receptor (SIGIRR), derived from recombinant SIGIRR (Met1~His118), is reactive with Human/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

The deduced 410-amino acid human protein, which is 82% identical to the mouse sequence and 23% identical to IL1R, has no signal peptide and contains a short 118-amino acid single immunoglobulin (Ig) extracellular domain with 4 potential N-glycosylation sites; a transmembrane region; and a 268-amino acid cytoplasmic tail with 4 IL1R-like signaling motifs. However, 2 changes in the regions conserved with IL1R (ser447 in IL1R replaced by cys222 in motif 3 of SIGIRR, and tyr536 in IL1R replaced by leu305 in motif 4 of SIGIRR) are predicted to abrogate signaling capabilities. Northern blot analysis revealed wide but differential expression of predominant 2.4- and 1.5-kb transcripts and minor 4.4- and 0.9-kb transcripts in all tissues tested.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: SIGIRR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Pig