- Antibody Type:Primary
- Antigen Name:Cannabinoid Receptor 1, Brain
- Antigen Symbol:CNR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:25248
- Antigen Synonyms:CANN6|CB1A|CNR|CB-R|CB1K5|CB1
- Amino Acid Number:1 - 117
- UniProtKB:P20272
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:17 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.46 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant CNR1 (Met1~Gln117)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAC950RA1
- Packaging:Vial
Polyclonal Antibody to Cannabinoid Receptor 1, Brain (CNR1), derived from recombinant CNR1 (Met1~Gln117), is reactive with Rat/Human/Mouse.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
The cannabinoid receptor type 1 is a G protein-coupled cannabinoid receptor that is found in the brain and is activated by the psychoactive drug cannabis and its active compound THC and by a group of endocannabinoid neurotransmitters including anandamide.The CB1 receptor is encoded by the gene or CNR1. Two transcript variants encoding different isoforms have been described for this gene., CNR1 orthologs have been identified in most mammals.CB1 receptors are thought to be the most widely expressed G protein-coupled receptors in the brain. The inverse agonist MK-9470 makes it possible to produce in vivo images of the distribution of CB1 receptors in the human brain with positron emission tomography.
Type: Primary
Antigen: CNR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat;Human;Mouse