- Antibody Type:Primary
- Antigen Name:Thrombospondin 4
- Antigen Symbol:THBS4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:TSP4
- Amino Acid Number:708 - 963
- UniProtKB:P49744
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:61 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.27 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant THBS4 (Pro708~Ile963)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAD824RA1
- Packaging:Vial
Polyclonal Antibody to Thrombospondin 4 (THBS4), derived from recombinant THBS4 (Pro708~Ile963), is reactive with Rat/Human/Mouse.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Thrombospondin 4 (THBS4) was originally identified from a Xenopus stage 45 cDNA library.Overall, the 961-amino acid predicted protein is 73% identical to the X. laevis sequence; however, the C-terminal 220 residues are over 92% identical. The TSP4 mRNA is approximately 3.4 kb. In contrast to thrombospondin-4 of Xenopus laevis, the human protein contains an RGD (arg-gly-asp) cell-binding sequence in the third type 3 repeat., Transfection of mouse NIH 3T3 fibroblasts or C2C12 myoblasts with a full-length human thrombospondin-4 cDNA resulted in the expression of a polypeptide with a reduced molecular weight of 140,000. In the absence of reducing agent, the expressed protein had an apparent molecular weight of 550000.
Type: Primary
Antigen: THBS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat;Human;Mouse