- Antibody Type:Primary
- Antigen Name:Sideroflexin 1
- Antigen Symbol:SFXN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:14057
- Antigen Synonyms:Tricarboxylate carrier protein|TCC
- Amino Acid Number:2 - 102
- UniProtKB:Q99JR1
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:15 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant SFXN1 (Ser2~Thr102)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAC843MU1
- Packaging:Vial
Polyclonal Antibody to Sideroflexin 1 (SFXN1), derived from recombinant SFXN1 (Ser2~Thr102), is reactive with Mouse/Human/Rat.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Recently sideroflexin 1 (Sfxn1) was shown to be a susceptible gene of flexed-tail (f/f) mice, and CTPL has turned out to be a rat orthologous protein of Sfxn3, a member of sideroflexin family. CTPL/Sfxn3 was targeted to mitochondrial membrane like Sfxn1., Might be involved in the transport of a component required for iron utilization into or out of the mitochondria. Recently sideroflexin 1 (Sfxn1) was shown to be a susceptible gene of flexed-tail (f/f) mice, and CTPL has turned out to be a rat orthologous protein of Sfxn3, a member of sideroflexinfamily.
Type: Primary
Antigen: SFXN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human;Rat