- Antibody Type:Primary
- Antigen Name:Thromboxane Receptor
- Antigen Symbol:TP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:6915
- Antigen Synonyms:Thromboxane A2 Receptor|Prostanoid TP receptor|TXA2-R|TBXA2R
- Amino Acid Number:3 - 324
- UniProtKB:P21731
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:36 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant TP (Pro3~Ser324)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAC822HU1
- Packaging:Vial
Polyclonal Antibody to Thromboxane Receptor (TP), derived from recombinant TP (Pro3~Ser324), is reactive with Human/Rat.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
The TP) is a protein on the surface of cells in the endothelium of blood vessels and in the placenta which interacts with the eicosanoid lipid thromboxane. It is a G-protein coupled receptors coupled to the G protein Gq.The gene responsible for the thromboxane receptor, TBXA2R is found on chromosome 19 and spans 15 kilobases., Thromboxane is a potent stimulator for platelet aggregation and clot formation and also plays a role in vascular tone. The thromboxane receptor is a member of the family of G protein-coupled receptors. Mutation of the receptor can lead to a bleeding disorder.
Type: Primary
Antigen: TP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat