Specifications
- Antibody Type:Primary
- Antigen Name:Von Willebrand Factor
- Antigen Symbol:vWF
- Clonality:Monoclonal
- Clone:C2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Rabbit
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:F8VWF|von Willebrand antigen 2|VWD
- UniProtKB:Q28834
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Storage Temperature:Store at 2...8 °C for frequent use, –20 ℃ for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C.
- Immunogen:Recombinant vWF
- Purification:Protein A + Protein G affinity chromatography
- Cat. No.:MSPP-MA833RB21
- Packaging:Vial
Specifications
About this item
Monoclonal Antibody to Von Willebrand Factor (vWF), derived from recombinant vWF, is reactive with Rabbit.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Von Willebrand factor (vWF) is a blood glycoprotein involved in hemostasis. It is deficient or defective in von Willebrand disease and is involved in a large number of other diseases, including thrombotic thrombocytopenic purpura, Heyde's syndrome, and possibly hemolytic-uremic syndrome. It is a large multimeric glycoprotein present in blood plasma and produced constitutively in endothelium (in the Weibel-Palade bodies), megakaryocytes (α-granules of platelets), and subendothelial connective tissue. Besides, it is not an enzyme and therefore has no catalytic activity. Its primary function is binding to other proteins, particularly Factor VIII and it is important in platelet adhesion to wound sites.
Type: Primary
Antigen: vWF
Clonality: Monoclonal
Clone: C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Rabbit