To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Interferon Inducible Protein 35
- Antigen Symbol:IFI35
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Pig,Human
- Western Blot:Yes
- Size:200 µl
- Environmentally Preferable:
- Cross Adsorption:No
- Form:Liquid
- Gene ID:3430
- Antigen Synonyms:IFP35
- Amino Acid Number:2 - 286
- UniProtKB:P80217
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:38 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, –20 ℃ for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C.
- Immunogen:Recombinant IFI35 (Ser2~Gly286)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAL605HU1
- Packaging:Vial
Polyclonal Antibody to Interferon Inducible Protein 35 (IFI35), derived from recombinant IFI35 (Ser2~Gly286), is reactive with Human/Pig.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
The cDNA encodes a 282-amino acid predicted protein containing a leucine zipper motif at the amino terminus. Unlike most other leucine zipper proteins, IFP35 lacks a basic domain associated with DNA binding. Northern blots showed that the 1.4-Kb IFP35 mRNA occurs in fibroblasts, macrophages, and epithelial cells following interferon treatment. Western blots indicated that the IFP35 gene product can form homodimers and that there was increased nuclear localization following interferon treatment. Among the exons identified from a phage P1 artificial chromosome (PAC) that included regions from BRCA1 were several coding regions with 100% identity to IFP35. IFP35 gene situated centromeric to the BRCA1 gene in the course of sequencing and analyzing 117 Kb of DNA from the 17q21 region.
Type: Primary
Antigen: IFI35
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Pig