- Antibody Type:Primary
- Antigen Name:Cefalexin
- Antigen Symbol:CEL
- Clonality:Monoclonal
- Clone:C6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Multiple
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:Cephalexin|Keflex
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Storage Temperature:Store at 2...8 °C for frequent use, –20 ℃ for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C.
- Immunogen:OVA conjugated CEL
- Purification:Protein A + Protein G affinity chromatography
- Cat. No.:MSPP-MK937GE21
- Packaging:Vial
Monoclonal Antibody to Cefalexin (CEL), derived from OVA conjugated CEL, is reactive with General species.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Cefalexin is used to treat a number of infections including: otitis media, streptococcal pharyngitis, bone and joint infections, pneumonia, cellulitis, and urinary tract infections. It may be used to prevent bacterial endocarditis. In addition to being a rational first-line treatment for cellulitis, it is a useful alternative to penicillins in patients with penicillin hypersensitivity. In patients with mild or questionable history of penicillin allergy, cephalasporins are now thought to be relatively safe. Caution should always be taken when prescribing cephalosporins to those with strong history of true penicillin hypersensitivity, however, because cefalexin and other first-generation cephalosporins are known to have a modest cross-allergy in patients with penicillin hypersensitivity. Cefalexin may not be effective against bacteria that incorporate a gene for beta-lactamase on their R-plasmid as the beta-lactam ring is present in the chemical structure of cephalosporin derivatives.
Type: Primary
Antigen: CEL
Clonality: Monoclonal
Clone: C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Multiple