Specifications
- Antibody Type:Primary
- Antigen Name:Interferon Tau
- Antigen Symbol:IFNt
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Bovine
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:317698
- Antigen Synonyms:Trophoblastin|Trophoblast Protein 1|Antiluteolysin|Interferon Tau-1|TP1|Trophoblast Antiluteolytic Protein|OTP|IFN-T1
- Amino Acid Number:24 - 195
- UniProtKB:P15696
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:23 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, –20 ℃ for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.98 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C.
- Immunogen:Recombinant IFNt (Cys24~Leu195)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAB862BO1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Interferon Tau (IFNt), derived from recombinant IFNt (Cys24~Leu195), is reactive with Bovine.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Interferon tau is a Type I interferon made up of a single chain of amino-acids. In many species of ruminant, it acts as a signaling molecule during pregnancy. It is secreted by the trophoblast cells into the uterine lumen in days 13 to 21 of pregnancy. It decreases endometrial oxytocin receptors, which then cannot stimulate PGF-2-alpha synthesis, preventing luteolysis. It promotes uterine implantation by increasing protein synthesis in glands. Although its role in humans is not certain, it has been found to bind to the same receptors sensitive to interferon alpha, and thus may have anti-viral properties, as hinted by studies showing its ability to inhibit the reverse transcriptase enzyme found in retroviruses such as HIV. Its low toxicity compared to other interferons also supports its investigation as a potential therapeutic compound.
Type: Primary
Antigen: IFNt
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine