Specifications
- Antibody Type:Primary
- Antigen Name:Apolipoprotein B
- Antigen Symbol:APOB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Pig,Human
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:338
- Antigen Synonyms:Apo B-100|Apo B-48|FLDB|Apo-B
- Amino Acid Number:3365 - 3548
- UniProtKB:P04114
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:26 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, –20 ℃ for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C.
- Immunogen:Recombinant APOB (His3365~Glu3548)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAC03HU01
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Apolipoprotein B (APOB), derived from recombinant APOB (His3365~Glu3548), is reactive with Human/Pig.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Apolipoprotein B (APOB) is the primary apolipoprotein of low-density lipoproteins (LDL or 'bad cholesterol'), which is responsible for carrying cholesterol to tissues. While it is unclear exactly what functional role APOB plays in LDL, it is the primary apolipoprotein component and is absolutely required for its formation. What is clear is that the APOB on the LDL particle acts as a ligand for LDL receptors in various cells throughout the body. Through a mechanism that is not fully understood, high levels of APOB can lead to plaques that cause vascular disease (atherosclerosis), leading to heart disease. There is considerable evidence that levels of APOB are a better indicator of heart disease risk than total cholesterol or LDL.
Type: Primary
Antigen: APOB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Pig