- Antibody Type:Primary
- Antigen Name:Estradiol
- Antigen Symbol:E2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Multiple
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:17B-Estradiol|Beta-Estradiol|Oestradiol
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:272.4 Da
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:OVA conjugated E2
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAA461GE1
- Packaging:Vial
Polyclonal Antibody to Estradiol (E2), derived from OVA conjugated E2, is reactive with General species.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Estradiol is an estrogenic hormone, C18H24O2, produced by the ovaries and used in treating estrogen deficiency. Estradiol is essential for maintaining normal female functions and is normally produced by the ovaries. After menopause, the ovaries decrease their production of this hormone.,Estradiol is a sex hormone. Estradiol is the predominant sex hormone present in females; however, it is present in males, albeit at lower levels, as well. It represents the major estrogen in humans. Estradiol has not only a critical impact on reproductive and sexual functioning, but also affects other organs including the bones.Estradiol is produced not only in the gonads: In both sexes, precursor hormones, to be specific testosterone, are converted by aromatization to estradiol. In particular, fat cells are active to convert precursors to estradiol, and will continue to do so even after menopause.
Type: Primary
Antigen: E2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: General species