Specifications
- Antibody Type:Primary
- Antigen Name:Fibrinogen Beta Chain
- Antigen Symbol:FGB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Pig,Human,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:2244
- Antigen Synonyms:Fibrinopeptide B|Fibrinogen Beta|FG-B
- Amino Acid Number:45 - 491
- UniProtKB:P02675
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:52 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant FGB(Gly45~Gln491)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAB215HU1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Fibrinogen Beta Chain (FGB), derived from recombinant FGB(Gly45~Gln491), is reactive with Human/Mouse/Pig.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Fibrinogen beta chain is found in humans and most other vertebrates with a similar system of blood coagulation. Fibrinogen beta chain is the beta component of fibrinogen, a blood-borne glycoprotein composed of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots.,In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in this gene lead to several disorders, including afibrinogenemia, dysfibrinogenemia, hypodysfibrinogenemia and thrombotic tendency.
Type: Primary
Antigen: FGB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Pig