ContactUsLinkComponent
Med Dimensions® Cockatoo Intubation Trainer
Med Dimensions® Cockatoo Intubation Trainer
  470357-262
New Product
undefined
Med Dimensions® Cockatoo Intubation Trainer
  470357-262
 :  MD-E-A1021S
New Product
  • Description:
    Cockatoo intubation trainer standard
  • Cat. No.:
    470357-262
Ready to take your avian veterinary skills to the next level?
  • Derived from CT scans, with relative anatomy including soft tongue, flexible jaw and airway to syrinx
  • Ideal for practicing intubation
  • Ideal for use with 3.5 ET tube
  • Bisected body to visualize the airway
Aspiring avian or exotics vets, this simulator is your key to acing cockatoo intubation. The Cockatoo Intubation Standard replicates the unique anatomy of a cockatoo's beak and airway. Build confidence by gaining hands-on experience before treating live birds. Improve skills by mastering essential intubation techniques in a safe environment. Enhance patient care by providing birds with the best possible care by honing your skills.
 : This product is a non-medical device intended solely for educational purposes. It is not designed, manufactured, or intended for use in the diagnosis, treatment, cure, mitigation, or prevention of disease or for any other medical purpose.