EnviroTrans™ Butterfield's Phosphate Buffer, Tube with Attached Swab, 1 ml
76492-666
:
- Pack type:20 per package
- Description:Butterfield's phosphate buffer, 15×75 mm
- Cat. No.:76492-666
- Fill volume:1 ml
Collect and transport your environmental samples from surfaces and transport to the lab efficiently and easily with EnviroTrans™.
- Flat bottom: Contributes to stability when set on end
- Leak-proof: Caps provide security during handling and transportation
- Attached swab: Collect and transport environmental samples with efficiency and ease
- Robust construction: Tubes have excellent impact and are shatter resistant
Butterfield's phosphate buffer is used as a diluent for the preparation of dilutions in plate count and other laboratory processes.
Collect and transport your samples with a few easy steps:
Swab surface and collect sample
Replace the cap and transport to lab
Sample can then be inoculated onto plated media for enumeration or used for pathogen detection
: 20 per package.