ContactUsLinkComponent
UMMt 2 ml Transport Medium
  76321-014
 :  
UMMt 2 ml Transport Medium
  76321-014
 :  00835
 :  
  • Description:
    UMMT, 2 ml, transport medium for mycofast
  • Size:
    35 tests
  • Cat. No.:
    76321-014
Transport medium for Mycofast, preserving specimen integrity for up to 72 hours before analysis, for Ureaplasma urealyticum (U.u.) and Mycoplasma hominis present in pathological patient samples.